accessibility menu, dialog, popup

UserName

H3CSNCH3ClN

CAS RN 17119-73-2

IUPAC Name: 4-chloro-6-methyl-2-(methylsulfanyl)pyrimidine
Synonyms: 4-chloro-6-methyl-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 174.65
Molecular Formula: C6H7ClN2S
InChi Key: ALMBOXQFPLQVLF-UHFFFAOYSA-N
SMILES: CSC1=NC(C)=CC(Cl)=N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results