accessibility menu, dialog, popup

UserName

CAS RN 17136-36-6

HOONH

CAS RN 17136-36-6

IUPAC Name: 2-(benzylamino)acetic acid
Synonyms: (benzylamino)acetic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: KGSVNOLLROCJQM-UHFFFAOYSA-N
SMILES: OC(=O)CNCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)