accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNCH3H3C

CAS RN 171364-78-6

IUPAC Name: N,N-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: N,N-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 247.15
Molecular Formula: C14H22BNO2
InChi Key: DGMLJJIUOFKPKB-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results