accessibility menu, dialog, popup

UserName

FFSFFS

CAS RN 1717-50-6

IUPAC Name: tetrafluoro-1,3-dithietane
Synonyms: tetrafluoro-1,3-dithietane
Molecular Weight (g/mol): 164.14
Molecular Formula: C2F4S2
InChi Key: MPOCEMBRPHOCMK-UHFFFAOYSA-N
SMILES: FC1(F)SC(F)(F)S1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results