accessibility menu, dialog, popup

UserName

CAS RN 17176-77-1

IUPAC Name: bis(benzyloxy)(oxo)-λ⁵-phosphanylium
Synonyms: bis(benzyloxy)(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 261.24
Molecular Formula: C14H14O3P
InChi Key: RQKYHDHLEMEVDR-UHFFFAOYSA-N
SMILES: O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)