CAS RN 17184-19-9
CAS RN 17184-19-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Desmethyl Deferiprone, TRC
CAS: 17184-19-9 Molecular Formula: C6H7NO2 Molecular Weight (g/mol): 125.13 Synonym: 2-Methyl-3-hydroxy-4(1H)-pyridone,2-Methyl-3-hydroxypyridine-4-one,3-Hydroxy-2-methyl-4(1H)-pyridone,CP 028,NSC 286640 SMILES: CC1=C(O)C(=O)C=CN1