accessibility menu, dialog, popup

UserName

CH3H3CNHNH2S

CAS RN 1719-76-2

IUPAC Name: (propan-2-yl)thiourea
Synonyms: urea, 1-isopropyl-2-thio-
Molecular Weight (g/mol): 118.2
Molecular Formula: C4H10N2S
InChi Key: POXAIQSXNOEQGM-UHFFFAOYSA-N
SMILES: CC(C)NC(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results