accessibility menu, dialog, popup

UserName

CAS RN 172967-04-3

IUPAC Name: 1-(2-fluoro-4-nitrophenyl)piperidine
Synonyms: 1-(2-fluoro-4-nitrophenyl)piperidine
Molecular Weight (g/mol): 224.24
Molecular Formula: C11H13FN2O2
InChi Key: MVYNWBRDGGKNCS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results