accessibility menu, dialog, popup

UserName

CH3NNH3C

CAS RN 1739-84-0

IUPAC Name: 1,2-dimethyl-1H-imidazole
Synonyms: 1H-imidazole, 1,2-dimethyl-
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: GIWQSPITLQVMSG-UHFFFAOYSA-N
SMILES: CN1C=CN=C1C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)