accessibility menu, dialog, popup

UserName

CH3NBOHHO

CAS RN 173999-18-3

IUPAC Name: (5-methylpyridin-3-yl)boronic acid
Synonyms: 5-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: REONQWGHSQHTAC-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results