accessibility menu, dialog, popup

UserName

CH3NSSH2N

CAS RN 174223-29-1

IUPAC Name: 2-methyl-1,3-thiazole-4-carbothioamide
Synonyms: 2-methyl-1,3-thiazole-4-carbothioamide
Molecular Weight (g/mol): 158.24
Molecular Formula: C5H6N2S2
InChi Key: DIOPPULTIGEDCB-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results