accessibility menu, dialog, popup

UserName

CAS RN 174855-53-9

IUPAC Name: tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 291.4
Molecular Formula: C16H25N3O2
InChi Key: UINNZXQKTNAOBL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results