accessibility menu, dialog, popup

UserName

CAS RN 17499-68-2

OHCOOCHOOCHH3COOHCOCH3ZrCH3H3CCF3CF3CF3CF3

CAS RN 17499-68-2

IUPAC Name: zirconium(4+) tetrakis(1,1,1-trifluoro-4-oxopent-2-en-2-olate)
Synonyms: zirconium(4+) tetrakis(1,1,1-trifluoro-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 703.55
Molecular Formula: C20H16F12O8Zr
InChi Key: WYYHZWGGPPBCMA-UHFFFAOYSA-J
SMILES: [Zr+4].CC(=O)C=C([O-])C(F)(F)F.CC(=O)C=C([O-])C(F)(F)F.CC(=O)C=C([O-])C(F)(F)F.CC(=O)C=C([O-])C(F)(F)F
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results