accessibility menu, dialog, popup

UserName

CH3OONNFFFCl

CAS RN 175137-27-6

IUPAC Name: methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Synonyms: methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 240.57
Molecular Formula: C7H4ClF3N2O2
InChi Key: VLUBJYUOPNGBOQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(Cl)N=C1C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results