accessibility menu, dialog, popup

UserName

CAS RN 175203-08-4

IUPAC Name: 6-(4-chlorophenyl)-1,2-dihydropyrimidine-2-thione
Synonyms: 4-(4-chlorophenyl)-3H-pyrimidine-2-thione
Molecular Weight (g/mol): 222.69
Molecular Formula: C10H7ClN2S
InChi Key: TWKIWOOKHPWUIM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=NC(=S)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results