CAS RN 175203-58-4
IUPAC Name:
4-(2-phenylethynyl)thiophene-2-carbaldehyde
Synonyms:
4-(2-phenylethynyl)thiophene-2-carbaldehyde
Molecular Weight (g/mol):
212.27
Molecular Formula:
C13H8OS
InChi Key:
SAYKNBQCVYQQJF-UHFFFAOYSA-N
SMILES:
O=CC1=CC(=CS1)C#CC1=CC=CC=C1
1
–
1
of
1
results
1
4-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde, 97%, Thermo Scientific™
CAS: 175203-58-4 Molecular Formula: C13H8OS Molecular Weight (g/mol): 212.266 MDL Number: MFCD00174359 InChI Key: SAYKNBQCVYQQJF-UHFFFAOYSA-N Synonym: 4-2-phenyleth-1-ynyl thiophene-2-carbaldehyde,4-2-phenylethynyl thiophene-2-carbaldehyde,4-phenylethynyl-thiophene-2-carbaldehyde,4-phenylethynyl thiophene-2-carbaldehyde,2-thiophenecarboxaldehyde, 4-phenylethynyl,4-phenylethynyl thiophene-2-carboxaldehyde PubChem CID: 2737139 IUPAC Name: 4-(2-phenylethynyl)thiophene-2-carbaldehyde SMILES: C1=CC=C(C=C1)C#CC2=CSC(=C2)C=O