accessibility menu, dialog, popup

UserName

CAS RN 175203-60-8

IUPAC Name: 2-bromo-5-chloro-3-methyl-1-benzothiophene
Synonyms: 2-bromo-5-chloro-3-methyl-1-benzothiophene
Molecular Weight (g/mol): 261.56
Molecular Formula: C9H6BrClS
InChi Key: RWFPSUKDYYBMHU-UHFFFAOYSA-N
SMILES: CC1=C(Br)SC2=CC=C(Cl)C=C12
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)