accessibility menu, dialog, popup

UserName

NONClSOOCl

CAS RN 175203-78-8

IUPAC Name: 5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride
Synonyms: 5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride
Molecular Weight (g/mol): 253.05
Molecular Formula: C6H2Cl2N2O3S
InChi Key: ZFBZICACMDZKNQ-UHFFFAOYSA-N
SMILES: ClC1=C(C2=NON=C2C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results