accessibility menu, dialog, popup

UserName

CH3SNNNH2NS

CAS RN 175204-61-2

IUPAC Name: 4-(methylsulfanyl)-6-(thiophen-2-yl)-1,3,5-triazin-2-amine
Synonyms: 4-(methylsulfanyl)-6-(thiophen-2-yl)-1,3,5-triazin-2-amine
Molecular Weight (g/mol): 224.3
Molecular Formula: C8H8N4S2
InChi Key: DKBHZJZOKUMDEF-UHFFFAOYSA-N
SMILES: CSC1=NC(=NC(N)=N1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results