accessibility menu, dialog, popup

UserName

CAS RN 175205-77-3

IUPAC Name: 4-chloro-2-(trifluoromethoxy)aniline
Synonyms: 4-chloro-2-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 211.57
Molecular Formula: C7H5ClF3NO
InChi Key: UCFTYLMKCJPWBX-UHFFFAOYSA-N
SMILES: NC1=C(OC(F)(F)F)C=C(Cl)C=C1
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)