accessibility menu, dialog, popup

UserName

CAS RN 175276-64-9

IUPAC Name: ethyl 5-(4-chlorophenyl)-2-formylfuran-3-carboxylate
Synonyms: ethyl 5-(4-chlorophenyl)-2-formylfuran-3-carboxylate
Molecular Weight (g/mol): 278.69
Molecular Formula: C14H11ClO4
InChi Key: XFOQMTOCKSVCBO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(OC(=C1)C1=CC=C(Cl)C=C1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results