accessibility menu, dialog, popup

UserName

ClHNNH2ClHCl

CAS RN 175276-76-3

IUPAC Name: 2-(2,6-dichlorophenyl)ethanimidamide hydrochloride
Synonyms: 2-(2,6-dichlorophenyl)ethanimidamide hydrochloride
Molecular Weight (g/mol): 239.52
Molecular Formula: C8H9Cl3N2
InChi Key: YNPZJFKKDYCULJ-UHFFFAOYSA-N
SMILES: Cl.NC(=N)CC1=C(Cl)C=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results