accessibility menu, dialog, popup

UserName

CH3OOCH3NH3CN

CAS RN 175277-33-5

IUPAC Name: 4-(dimethoxymethyl)-2-methylpyrimidine
Synonyms: 4-(dimethoxymethyl)-2-methylpyrimidine
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: VDXHJWMLYFVGDL-UHFFFAOYSA-N
SMILES: COC(OC)C1=CC=NC(C)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results