accessibility menu, dialog, popup

UserName

FFFSClOO

CAS RN 175278-08-7

IUPAC Name: 2,3,4-trifluorobenzene-1-sulfonyl chloride
Synonyms: 2,3,4-trifluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 230.59
Molecular Formula: C6H2ClF3O2S
InChi Key: XFTDZYFHXRZLEF-UHFFFAOYSA-N
SMILES: FC1=CC=C(C(F)=C1F)S(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)