accessibility menu, dialog, popup

UserName

NONHO

CAS RN 175609-19-5

IUPAC Name: (2,1,3-benzoxadiazol-4-yl)methanol
Synonyms: 2,1,3-benzoxadiazol-4-ylmethanol
Molecular Weight (g/mol): 150.14
Molecular Formula: C7H6N2O2
InChi Key: OKDMFOBEJCALFD-UHFFFAOYSA-N
SMILES: OCC1=CC=CC2=NON=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results