accessibility menu, dialog, popup

UserName

H3NO

CAS RN 1758-46-9

IUPAC Name: 2-phenoxyethan-1-aminium
Synonyms: 2-phenoxyethanaminium
Molecular Weight (g/mol): 138.19
Molecular Formula: C8H12NO
InChi Key: IMLAIXAZMVDRGA-UHFFFAOYSA-O
SMILES: [NH3+]CCOC1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)