accessibility menu, dialog, popup

UserName

CAS RN 177-11-7

IUPAC Name: 1,4-dioxa-8-azaspiro[4.5]decan-8-ium
Synonyms: 1,4-dioxa-8-azaspiro[4.5]decan-8-ium
Molecular Weight (g/mol): 144.19
Molecular Formula: C7H14NO2
InChi Key: KPKNTUUIEVXMOH-UHFFFAOYSA-O
SMILES: C1COC2(CC[NH2+]CC2)O1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)