accessibility menu, dialog, popup

UserName

H2NNONH(E/Z)

CAS RN 1772-01-6

IUPAC Name: 1-(1,2-dihydropyridin-2-ylidene)-1-nitrosomethanamine
Synonyms: 1-nitroso-1-(1H-pyridin-2-ylidene)methanamine
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: NJNUYQDYGLUAAG-UHFFFAOYSA-N
SMILES: NC(N=O)=C1NC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results