accessibility menu, dialog, popup

UserName

HOFF

CAS RN 178312-48-6

IUPAC Name: (4,4-difluorocyclohexyl)methanol
Synonyms: (4,4-difluorocyclohexyl)methanol
Molecular Weight (g/mol): 150.17
Molecular Formula: C7H12F2O
InChi Key: XJZNZSLOHZLFQP-UHFFFAOYSA-N
SMILES: OCC1CCC(F)(F)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results