accessibility menu, dialog, popup

UserName

CAS RN 179067-99-3

CH3OONHOO(E/Z)

CAS RN 179067-99-3

IUPAC Name: ethyl 6-(hydroxyimino)-2-oxatricyclo[5.4.0.0³,⁵]undeca-1(11),7,9-triene-3-carboxylate
Synonyms: cpccoet
Molecular Weight (g/mol): 247.25
Molecular Formula: C13H13NO4
InChi Key: FXCTZFMSAHZQTR-UHFFFAOYNA-N
SMILES: CCOC(=O)C12CC1C(=NO)C1=CC=CC=C1O2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results