accessibility menu, dialog, popup

UserName

OHBHOOFFF

CAS RN 179113-90-7

IUPAC Name: [3-(trifluoromethoxy)phenyl]boronic acid
Synonyms: 3-(trifluoromethoxy)phenylboronic acid
Molecular Weight (g/mol): 205.93
Molecular Formula: C7H6BF3O3
InChi Key: UWDFWVLAHRQSKK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(OC(F)(F)F)=C1
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results