accessibility menu, dialog, popup

UserName

CAS RN 179250-28-3

IUPAC Name: tert-butyl 4-[2-(hydroxymethyl)phenyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[2-(hydroxymethyl)phenyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 292.38
Molecular Formula: C16H24N2O3
InChi Key: MQZUQGHXDLMAKT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1CO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results