accessibility menu, dialog, popup

UserName

AlAlOSOOOOSOOOOSOOO

CAS RN 17927-65-0

IUPAC Name: dialuminium(3+) hydrate trisulfate
Synonyms: dialuminium(3+) hydrate trisulfate
Molecular Weight (g/mol): 360.15
Molecular Formula: Al2H2O13S3
InChi Key: BUACSMWVFUNQET-UHFFFAOYSA-H
SMILES: O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (9)
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
Grade 
  • (2)
  • (3)
Physical Form 
  • (5)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (9)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)