CAS RN 179333-18-7
CAS RN 179333-18-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
NGD 94-1, TRC
CAS: 179333-18-7 Molecular Formula: C18 H20 N6 . 2[C4 H4 O4] Molecular Weight (g/mol): 552.536 Synonym: NGD 94-1,CPDD 0045,NGD 941 IUPAC Name: (Z)-but-2-enedioic acid;2-[4-[(2-phenyl-1H-imidazol-5-yl)methyl]piperazin-1-yl]pyrimidine SMILES: OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O.C(N1CCN(CC1)c2ncccn2)c3cnc([nH]3)c4ccccc4