CAS RN 1793944-59-8
CAS RN 1793944-59-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pyronil 45-d34 (>90%), TRC
CAS: 1793944-59-8 Molecular Formula: C24D34Br4O4 Molecular Weight (g/mol): 740.35 Synonym: 3,4,5,6-Tetrabromo-1,2-benzenedicarboxylic Acid 1,2-Bis(2-ethylhexyl-d34) Ester,Bis(2-ethylhexyl-d34) Tetrabromophthalate,DP 45,Di(2-ethylhexyl-34) Tetrabromophthalate,Uniplex FRP 45-d34,TBPH-d34 SMILES: BrC1=C(Br)C(C(OC([2H])([2H])C(C([2H])(C([2H])([2H])[2H])[2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)=C(C(OC([2H])([2H])C(C([2H])([2H])C([2H])([2H])[2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)C(Br)=C1Br