CAS RN 1797983-48-2
CAS RN 1797983-48-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lamotrigine 3,3-Dimer, TRC
CAS: 1797983-48-2 Molecular Formula: C19 H14 Cl4 N10 Molecular Weight (g/mol): 524.19 Synonym: N3-[[[5-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]amino]methyl]-6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine IUPAC Name: 3-N-[[[5-amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]amino]methyl]-6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine SMILES: Nc1nc(NCNc2nnc(c(N)n2)c3cccc(Cl)c3Cl)nnc1c4cccc(Cl)c4Cl