CAS RN 180028-20-0
CAS RN 180028-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dihydroxybenzaldehyde-d3, TRC
CAS: 180028-20-0 Molecular Formula: C7H3D3O3 Molecular Weight (g/mol): 141.14 Synonym: 4,5-Dihydroxybenzaldehyde-2,3,6-D3 IUPAC Name: 2,3,6-trideuterio-4,5-dihydroxybenzaldehyde SMILES: [2H]c1c([2H])c(C=O)c([2H])c(O)c1O