accessibility menu, dialog, popup

UserName

CAS RN 18055-70-4

CH3SiH3CCH3OSiH3CCH3H3C

CAS RN 18055-70-4

IUPAC Name: {[dimethyl(4-methylphenyl)silyl]oxy}dimethyl(4-methylphenyl)silane
Synonyms: {[dimethyl(4-methylphenyl)silyl]oxy}dimethyl(4-methylphenyl)silane
Molecular Weight (g/mol): 314.58
Molecular Formula: C18H26OSi2
InChi Key: MEQYEVSZYLVDNX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)[Si](C)(C)O[Si](C)(C)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results