accessibility menu, dialog, popup

UserName

OOMgBr

CAS RN 180675-22-3

IUPAC Name: magnesium(2+) (1,3-dioxolan-2-yl)methanide bromide
Synonyms: magnesium(2+) 1,3-dioxolan-2-ylmethanide bromide
Molecular Weight (g/mol): 191.31
Molecular Formula: C4H7BrMgO2
InChi Key: IERSPCAGKAOLSQ-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].[CH2-]C1OCCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)