CAS RN 1808-26-0
CAS RN 1808-26-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl Arachidonate, TRC
CAS: 1808-26-0 Molecular Formula: C22 H36 O2 Molecular Weight (g/mol): 332.52 Synonym: (5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic Acid Ethyl Ester,(all-Z)-5,8,11,14-Eicosatetraenoic Acid Ethyl Ester,Arachidonic Acid Ethyl Ester IUPAC Name: ethyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate SMILES: CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OCC