accessibility menu, dialog, popup

UserName

LiCH3OCH3O(E/Z)

CAS RN 18115-70-3

IUPAC Name: lithium(1+) 2,4-dioxopentan-3-ide
Synonyms: lithium(1+) acetylacetonate
Molecular Weight (g/mol): 106.05
Molecular Formula: C5H7LiO2
InChi Key: YRDZXTGCHNVCJJ-UHFFFAOYSA-N
SMILES: [Li+].CC(=O)[CH-]C(C)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results