CAS RN 18125-49-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxymethylantipyrine, TRC
CAS: 18125-49-0 Molecular Formula: C11H12N2O2 Molecular Weight (g/mol): 204.23 Synonym: 3-(Hydroxymethyl)-2-methyl-1-phenyl-3-pyrazolin-5-one,HMA,1,2-Dihydro-5-(hydroxymethyl)-1-methyl-2-phenyl-3H-pyrazol-3-one,3-(Hydroxymethyl)-2-methyl-1-phenyl-3-pyrazolin-5-one,Hydroxymethylantipyrine IUPAC Name: 5-(hydroxymethyl)-1-methyl-2-phenylpyrazol-3-one SMILES: CN1N(C(=O)C=C1CO)c2ccccc2