CAS RN 18163-47-8
IUPAC Name:
(2-iodoethynyl)trimethylsilane
Synonyms:
(2-iodoethynyl)trimethylsilane
Molecular Weight (g/mol):
224.12
Molecular Formula:
C5H9ISi
InChi Key:
HNIRHTRSZDSMOF-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C#CI
1
–
1
of
1
results
1
1-Iodo-2-(trimethylsilyl)acetylene, 97%
CAS: 18163-47-8 Molecular Formula: C5H9ISi Molecular Weight (g/mol): 224.12 MDL Number: MFCD00274201 InChI Key: HNIRHTRSZDSMOF-UHFFFAOYSA-N Synonym: 1-iodo-2-trimethylsilyl acetylene,iodoethynyl trimethylsilane,silane, iodomethyl trimethyl,2-iodoethynyl trimethylsilane,2-iodoethynyl trimethyl silane,silane, iodoethynyl trimethyl,tbdmcs,acmc-20aoug,trimethyliodoethinylsilane,iodoethynyltrimethylsilane PubChem CID: 140341 IUPAC Name: 2-iodoethynyl(trimethyl)silane SMILES: C[Si](C)(C)C#CI