CAS RN 18197-26-7
CAS RN 18197-26-7
IUPAC Name:
sodium diformylazanide
Synonyms:
sodium diformylazanide
Molecular Weight (g/mol):
95.03
Molecular Formula:
C2H2NNaO2
InChi Key:
QJXDSDLNUKLDBP-UHFFFAOYSA-M
SMILES:
[Na+].O=C[N-]C=O
1
–
1
of
1
results
1
Sodium diformamide, 98%
CAS: 18197-26-7 Molecular Formula: C2H2NNaO2 Molecular Weight (g/mol): 95.03 MDL Number: MFCD00145302 InChI Key: QJXDSDLNUKLDBP-UHFFFAOYSA-M Synonym: sodium diformylamide,diformylamide sodium salt,diformylimide sodium salt,sodium diformylazanide,sodium diformamide,diformylamino sodium,diformylsodium,acmc-209eii,sodium dimethanoylazanide,ksc538a2h PubChem CID: 10877056 IUPAC Name: sodium;diformylazanide SMILES: C(=O)[N-]C=O.[Na+]