accessibility menu, dialog, popup

UserName

H3COOSNNH3C

CAS RN 18212-20-9

IUPAC Name: ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate
Synonyms: ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: AHPXTXGCMLOXGA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=NS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results