accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHNSNOHNH2(E/Z)

CAS RN 1824739-91-4

IUPAC Name: tert-butyl N-{4-[4-(N'-hydroxycarbamimidoyl)phenyl]-1,3-thiazol-2-yl}carbamate
Synonyms: tert-butyl N-{4-[4-(N'-hydroxycarbamimidoyl)phenyl]-1,3-thiazol-2-yl}carbamate
Molecular Weight (g/mol): 334.39
Molecular Formula: C15H18N4O3S
InChi Key: UERHOGGYEPCACK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC(=CS1)C1=CC=C(C=C1)C(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results