accessibility menu, dialog, popup

UserName

NOCl

CAS RN 182691-76-5

IUPAC Name: 5-chlorofuro[3,2-b]pyridine
Synonyms: 5-chlorofuro[3,2-b]pyridine
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: LBJDELVHPFXIRO-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results