accessibility menu, dialog, popup

UserName

ICH3SH3CH3C

CAS RN 1829-92-1

IUPAC Name: triethylsulfanium iodide
Synonyms: triethylsulfanium iodide
Molecular Weight (g/mol): 246.15
Molecular Formula: C6H15IS
InChi Key: LKDQWVKWYGOVJW-UHFFFAOYSA-M
SMILES: [I-].CC[S+](CC)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results