CAS RN 183162-43-8
IUPAC Name:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol
Molecular Weight (g/mol):
562.15
Molecular Formula:
C12H6F20O2
InChi Key:
MBKZIVRAOJBYDI-UHFFFAOYSA-N
SMILES:
OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
1
–
1
of
1
results
1
1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, tech. 90%, Thermo Scientific Chemicals
CAS: 183162-43-8 Molecular Formula: C12H6F20O2 Molecular Weight (g/mol): 562.146 MDL Number: MFCD00236625 InChI Key: MBKZIVRAOJBYDI-UHFFFAOYSA-N Synonym: 1h,1h,12h,12h-perfluoro-1,12-dodecanediol,1h,1h,12h,12h-perfluorododecanediol,1h,1h,12h,12h-perfluoro-1,12-dodecandiol,1h,1h,12h,12h-perfluorododecane-1,12-diol,1h,1h,12h,12h-icosafluoro-1,12-dodecanediol,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosakis fluoranyl dodecane-1,12-diol PubChem CID: 2776100 IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluorododecane-1,12-diol SMILES: C(C(C(C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O