accessibility menu, dialog, popup

UserName

CH3ClN

CAS RN 18436-73-2

IUPAC Name: 4-chloro-8-methylquinoline
Synonyms: 4-chloro-8-methylquinoline
Molecular Weight (g/mol): 177.63
Molecular Formula: C10H8ClN
InChi Key: PGDPMZFATHZAIQ-UHFFFAOYSA-N
SMILES: CC1=C2N=CC=C(Cl)C2=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results